EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Spectrophotometric determination of cobalt(II) and cyanocobalamin with vanillilfluorone and its applications.
PMID 21628907
Abstract
Spectrophotometric determination of cobalt(II) was accomplished with vanillilfluorone (VF) in the presence of dimethylbenzyltetradecylammonium chloride (Zephiramine, Zep). In the determination of cobalt(II), Beer's law was obeyed in the range of 24-470 ng/ml, with an effective molar absorption coefficient (at 575 nm) and relative standard deviation of 1.35×10(5) l mol(-1) cm(-1) and 0.66% (n=5), respectively. The composition ratio of the colored complex was determined by the mole ratio and continuous variation methods, and it was found to be Co(II) : VF : Zep=1 : 2 : 4. Analysis of cyanocobalamin by the same procedure showed that cyanocobalamin could be determined in the concentration range of 0.5-0.11 µg/ml using the proposed method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
13401
Benzyldimethyltetradecylammonium chloride, anhydrous, ≥99.0% (AT)
C23H42ClN