EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Selective visualization of point defects in carbon nanotubes at the atomic scale by an electron-donating molecular tip.
PMID 21629907
Abstract
Electron-donating molecular tips were used for the observation of single-walled carbon nanotubes (SWNTs). Defects in SWNTs were selectively visualized at the atomic scale on the basis of charge-transfer interaction with the molecular tip.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
422967
4-Aminothiophenol, 97%
C6H7NS