EMAIL THIS PAGE TO A FRIEND
Ecotoxicology (London, England)
Are water vole resistant to anticoagulant rodenticides following field treatments?
PMID 21630005
Abstract
The anti-vitamin Ks (AVKs) are widely used to control rodent populations. They inhibit Vitamin K regeneration by the Vitamin K Epoxide Reductase (VKOR) and cause a fatal hemorrhagic syndrome. Because of repeated use, some populations of commensal rodents have expressed resistance to these compounds. In Franche-Comté (France), the water vole exhibits cyclic population outbreaks. A second generation AVK, bromadiolone, has been used for the last 20 years to control vole populations. The aim of this study is to determine whether these repeated treatments could have led to the development of resistance to AVKs in water vole populations. We conducted enzymatic and genetic studies on water voles trapped in treated and non treated plot. The results indicate that voles from the most heavily treated area exhibit enzymatic changes in VKOR activity hence arguing for resistance to AVKs and that an intronic haplotype on the vkorc1 gene seems to be associated with these enzymatic changes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46035
Bromadiolon, PESTANAL®, analytical standard
C30H23BrO4