EMAIL THIS PAGE TO A FRIEND
Journal of veterinary emergency and critical care (San Antonio, Tex. : 2001)
4-Methylpyrazole as a treatment in naturally occurring ethylene glycol intoxication in cats.
PMID 21631713
Abstract
To describe the clinical experience and therapeutic use of fomepizole (4-methylpyrazole [4-MP]) in 3 cats with naturally occurring ethylene glycol (EG) toxicity. All cats were documented to be EG positive by an ethylene glycol test kit. This report describes the dose of 4-MP used, available clinicopathological data, and clinical progression during hospitalization. All patients survived to discharge. IV use of 4-MP at 125 mg/kg as an initial dose and 31.25 mg/kg at 12, 24, and 36 hours is safe and effective for treatment of naturally occurring EG toxicity in cats. Increased HCO₃ concentrations were noted after IV use of 4-MP. This is the first report documenting the successful treatment of naturally occurring EG intoxication in cats with 4-MP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1387
4-Methylpyrazole hydrochloride, alcohol dehydrogenase inhibitor
C4H6N2 · HCl