EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
Reutericyclin and related analogues kill stationary phase Clostridium difficile at achievable colonic concentrations.
PMID 21632577
Abstract
The stationary phase of Clostridium difficile, which is associated with the symptoms of the diarrhoeal disease, is refractory to antibiotic killing. The aim of this study was to explore whether probiotic-derived reutericyclin and related synthetic analogues could kill stationary phase C. difficile at concentrations achievable in the gastrointestinal tract. The bactericidal activities of reutericyclin and lead compound derivatives were examined against logarithmic and stationary phase cultures of different C. difficile strains. The absorption of compounds across the intestinal epithelia was tested using the Caco-2 permeability model. Unlike vancomycin and metronidazole, reutericyclins demonstrated concentration-dependent killing, being rapidly bactericidal against both logarithmic and stationary phase cells, at low concentrations (0.09-2 mg/L). The intestinal absorption of unmodified reutericyclin was poor and comparable to that of vancomycin. However, this property varied significantly for the synthetic reutericyclin analogues, ranging from well absorbed to non-absorbed. The non-absorbable compounds were highly effluxed, suggesting this parameter could be modulated to obtain agents with superior efficacy. Reutericyclins showed excellent potency against the lethal non-growing stage of C. difficile at concentrations that may be attained in the gastrointestinal tract. Since these agents represent novel potential treatments for C. difficile infection, further development of this compound class is warranted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37018
Tenuazonic acid, analytical standard
C10H15NO3
T3408
Tenuazonic acid copper salt from Alternaria alternata
C10H14NO3 · 0.5Cu