EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Racemase activity of B. cepacia lipase leads to dual-function asymmetric dynamic kinetic resolution of α-aminonitriles.
PMID 21633990