EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Substituted benzoxazinones as potent positive allosteric AMPA receptor modulators: part II.
PMID 21636273
Abstract
AMPA receptors (AMPARs) are an important therapeutic target in the CNS. A series of substituted benzoxazinone derivatives with good to very good in vitro activity as positive allosteric AMPAR modulators was synthesized and evaluated. The appropriate substituent choice on the benzoxazinone fragment improved the affinity towards the AMPA receptor significantly in comparison to our lead molecule CX614.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9950
Aniracetam, ≥98%
C12H13NO3