EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Pyridazine-derived γ-secretase modulators.
PMID 21636276
Abstract
SAR of a novel series of pyridazine-derived γ-secretase modulators is described. Compound 25 was found to be a potent modulator in vitro, which on further profiling, was found to decrease Aβ42 and Aβ40, and maintain the levels of total Aβ. Furthermore, 25 demonstrated excellent pharmacokinetic parameters as well as good CNS penetration in the rat.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P57204
Pyridazine, 98%
C4H4N2