EMAIL THIS PAGE TO A FRIEND
Acta odontologica latinoamericana : AOL
A preliminary study of enamel remineralization by dentifrices based on Recalden (CPP-ACP) and Novamin (calcium-sodium-phosphosilicate).
PMID 21638965
Abstract
The purpose of this study was to investigate the enamel remineralization potential of two toothpastes, one of which was based on Recaldent (CPP- ACP) and the other on NovaMin (Calcium-sodium-phosphosilicate). Human permanent molar teeth were subjected to three consecutive demineralization cycles. These cycles were followed by remineralization of the experimental groups by toothpastes containing Recalden and NovaMin respectively. The samples were analyzed by Scanning Electron Microscope, (SEM) and energy-dispersive X-ray spectroscopy analysis (EDX). Extensive demineralization was noted in the control group (without remineralization) while the groups treated with the dentifices demonstrated various degrees of remineralization, as shown by formation of different types of deposits on the enamel surface. The EDX analysis showed increased amounts of Ca, P Si and Zn in the enamel of the experimental groups, compared to the control one. Toothpastes containing Recalden and especially NovaMin have the potential to remineralize enamel, a property which might be important in finding a substitute to pit and fissure sealing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
71746
Sodium metasilicate pentahydrate, ≥95.0% (T)
Na2O3Si · 5H2O