EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
A general copper-BINAP-catalyzed asymmetric propargylation of ketones with propargyl boronates.
PMID 21639096
Abstract
An operationally simple copper-BINAP-catalyzed, highly enantioselective propargylation of ketones is presented. The methodology was developed as an enantioselective process for methyl ethyl ketone and shown to be applicable to a wide variety of prochiral ketones. The resulting homopropargyl adducts are versatile latent carbonyls from which γ-butyrolactones, β-hydroxy methyl ketones, and β-hydroxycarboxylates are readily obtained.