EMAIL THIS PAGE TO A FRIEND
Biochemistry. Biokhimiia
Three-dimensional structures of noncovalent complexes of Citrobacter freundii methionine γ-lyase with substrates.
PMID 21639836
Abstract
Crystal structures of Citrobacter freundii methionine γ-lyase complexes with the substrates of γ- (L-1-amino-3-methylthiopropylphosphinic acid) and β- (S-ethyl-L-cysteine) elimination reactions and the competitive inhibitor L-norleucine have been determined at 1.45, 1.8, and 1.63 Å resolution, respectively. All three amino acids occupy the active site of the enzyme but do not form a covalent bond with pyridoxal 5'-phosphate. Hydrophobic interactions between the active site residues and the side groups of the substrates and the inhibitor are supposed to cause noncovalent binding. Arg374 and Ser339 are involved in the binding of carboxyl groups of the substrates and the inhibitor. The hydroxyl of Tyr113 is a potential acceptor of a proton from the amino groups of the amino acids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C121908
L-Cysteine ethyl ester hydrochloride, 98%
C5H11NO2S · HCl