EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
In vivo invasion of head and neck squamous cell carcinoma cells does not require macrophages.
PMID 21641405
Abstract
Invasion of tumor cells into the local stroma is an important component in cancer progression. Here we report studies of the in vivo invasion of head and neck squamous cell carcinoma (HNSCC) cells in response to applied gradients of a growth factor [epidermal growth factor (EGF)] and a chemokine (CXCL12), using orthotopic floor-of-mouth models. Analysis of the invading cells indicated that >75% of them were tumor cells, about 15% macrophages, and <10% were unidentified. Surprisingly, although macrophages invaded together with tumor cells, macrophage contributions were not required for HNSCC invasion. CXCL12-induced in vivo invasion of HNSCC cells was also observed and found to occur via a unidirectional transactivation of epidermal growth factor receptor (EGFR) through CXCR4. Inhibition of tumor necrosis factor-α-converting enzyme using TNF-α protease inhibitor-2 selectively inhibited CXCL12-induced invasion but not EGF-induced invasion, consistent with CXCL12 activation of EGFR via release of EGFR ligands.