EMAIL THIS PAGE TO A FRIEND
The Medical journal of Australia
Extensively drug-resistant tuberculosis: New Zealand's first case and the challenges of management in a low-prevalence country.
PMID 21644877
Abstract
In 2010, an immigrant from Burma was the first person to be diagnosed in New Zealand with extensively drug-resistant tuberculosis (XDR-TB). The strain of Mycobacterium tuberculosis is the most resistant reported to date in Australasia. Key difficulties of managing this disease in a low-prevalence country were delays from drug-susceptibility testing and in acquiring appropriate medicines, and a lack of evidence-based guidelines. Solutions are needed for New Zealand and the wider region as more cases of XDR-TB are likely to be encountered in the future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09415
Sodium 4-aminosalicylate dihydrate, ≥97.0% (T)
C7H6NNaO3 · 2H2O