EMAIL THIS PAGE TO A FRIEND
Immunology letters
Upregulation of heme oxygenase-1 via PI3K/Akt and Nrf-2 signaling pathways mediates the anti-inflammatory activity of Schisandrin in Porphyromonas gingivalis LPS-stimulated macrophages.
PMID 21645546
Abstract
The lipopolysaccharide (LPS) of Porphyromonas gingivalis is thought to induce periodontitis. In this study, we isolated Schisandrin from the dried fruits of Schisandra chinensis and examined the anti-inflammatory effect of Schisandrin in macrophages stimulated with LPS from P. gingivalis. First, Schisandrin inhibited LPS-induced pro-inflammatory cytokines, including TNF-α, IL-1β, and IL-6. And Schisandrin suppressed the nuclear translocation and activity of NF-κB and phosphorylation of IκBα in LPS-stimulated RAW 264.7 cells. Next, the presence of a selective inhibitor of HO-1 (SnPP) and a siRNA specific for HO-1 inhibited Schisandrin-mediated anti-inflammatory activity. Furthermore, Schisandrin induced HO-1 expression of RAW 264.7 cells through Nrf-2, PI3K/Akt, and ERK activation. Therefore, these results suggest that the anti-inflammatory effects of Schisandrin on P. gingivalis LPS-stimulated RAW 264.7 cells may be due to a reduction of NF-κB activity and induction of the expression of HO-1, leading to TNF-α, IL-1β, and IL-6 down-regulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0054
Schizandrin, ≥98% (HPLC)
C24H32O7