EMAIL THIS PAGE TO A FRIEND
Talanta
Comparative study of the adsorption performance of an active multi-sorbent bed tube (Carbotrap, Carbopack X, Carboxen 569) and a Radiello(®) diffusive sampler for the analysis of VOCs.
PMID 21645756
Abstract
A simple comparison is made to evaluate the relative performance of active and passive sampling methods for the analysis of volatile organic compounds (VOCs) in ambient air. The active sampling is done through a multi-sorbent bed tube (Carbotrap, Carbopack X, Carboxen 569) created in our laboratory and the passive sampling through the Radiello(®) diffusive sampler specified for thermal desorption (filled with Carbograph 4). Daily duplicate samples of multi-sorbent bed tubes were taken during a period of 14 days. During the same period of time, quadruplicate samples of Radiello(®) tubes were taken during 3 days, 4 days, 7 days and 14 days. The sampling was carried out indoors during the months of February and March 2010 and outdoors during the month of July 2010 in La Canonja (Tarragona, Spain). The analysis was performed by automatic thermal desorption (ATD) coupled with capillary gas chromatography (GC)/mass spectrometry detector (MSD). The analytical performance of the two sampling approaches was evaluated by describing several quality assurance parameters. The results show that the analytical performances of the methodologies studied are quite similar. They display low limits of detection, good precision, accuracy and desorption efficiency, and low levels of breakthrough for multi-sorbent bed tubes. However, the two monitoring methods produced varying air-borne concentration data for most of the studied compounds, and the Radiello(®) samplers generally gave higher results. Sampling rates (Q(k)) were determined experimentally, and their values were higher than those supplied by the producer. As the experimental calculation of Q(k) values is generally carried out by the suppliers in exposure chambers with only the target compounds present in the air samples, as well as in concentrations dissimilar to those found in ambient air, the use of constant settled Q(k) can lead to inaccurate results in complex samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
25087
Carbotrap® 100, glass TD tube, preconditioned
20242 Carbotrap® 200 Dynatherm 890, 850, for use with Dynatherm ACEM-9300, ACEM-900 850/890 TDU, MTDU and OI Analytical Air Tube Desorber or DMP-16
20865 Carbotrap® 201, for use with Dynatherm ACEM 900/901-FF, I.D. × O.D. × L 1 mm × 6 mm × 7.25 in.
20083-U
Carbotrap® 202, stainless steel TD tube, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
29745-U
Carbotrap® 202, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20013-U
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
29531-U Carbotrap® 217, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
30138-U Carbotrap® 217, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2 , unconditioned, pkg of 10 ea
30137-U
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4 × 3 1/2 , unconditioned, pkg of 10 ea
29744-U
Carbotrap® 217, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29750-U Carbotrap® 217, O.D. × L 1/4  in. × 3 1/2  in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20875
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 4.5 in., preconditioned, pkg of 1 ea
28283-U
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
28273-U
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
20370-U Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 1/4  in. × 5 mm × 7 in., preconditioned, pkg of 1 ea
21705-U
Carbotrap® 300, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, preconditioned, pkg of 10 ea
29532-U
Carbotrap® 300, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
30141-U
Carbotrap® 300, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., unconditioned, pkg of 10 ea
30139-U
Carbotrap® 300, stainless steel TD tube, unconditioned
29746-U
Carbotrap® 300, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29751-U
Carbotrap® 300, O.D. × L 1/4  in. × 3 1/2  in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20881 Carbotrap® 317, glass TD tube (fast-flow), preconditioned
28311-U Carbotrap® 349, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
29752-U Carbotrap® 349, O.D. × L 1/4  in. × 3 1/2  in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20287 Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, bottle of 10 g
20309 Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 10 g
11047-U Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 500 g
10435-U Carbotrap® Adsorbent, matrix Carbotrap® X, 20-40 mesh, bottle of 10 g
10460-U Carbotrap® Adsorbent, matrix Carbotrap® Y, 20-40 mesh, bottle of 10 g
11325-U Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, pack of 144 × 190 mg