EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Removal of reactive red-120 and 4-(2-pyridylazo) resorcinol from aqueous samples by Fe3O4 magnetic nanoparticles using ionic liquid as modifier.
PMID 21645968
Abstract
The nanoparticles of Fe(3)O(4) as well as the binary nanoparticles of ionic liquid and Fe(3)O(4) (IL-Fe(3)O(4)) were synthesized for removal of reactive red 120 (RR-120) and 4-(2-pyridylazo) resorcinol (PAR) as model azo dyes from aqueous solutions. The mean size and the surface morphology of the nanoparticles were characterized by TEM, DLS, XRD, FTIR and TGA techniques. Adsorption of RR-120 and PAR was studied in a batch reactor at different experimental conditions such as nanoparticle dosage, dye concentration, pH of the solution, ionic strength, and contact time. Experimental results indicated that the IL-Fe(3)O(4) nanoparticles had removed more than 98% of both dyes under the optimum operational conditions of a dosage of 60mg, a pH of 2.5, and a contact time of 2min when initial dyes concentrations of 10-200mg L(-1) were used. The maximum adsorption capacity of IL-Fe(3)O(4) was 166.67 and 49.26mg g(-1) for RR-120 and PAR, respectively. The isotherm experiments revealed that the Langmuir model attained better fits to the equilibrium data than the Freundlich model. The Langmuir adsorption constants were 5.99 and 3.62L mg(-1) for adsorptions of RR-120 and PAR, respectively. Both adsorption processes were endothermic and dyes could be desorbed from IL-Fe(3)O(4) by using a mixed NaCl-acetone solution and adsorbent was reusable.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323209
4-(2-Pyridylazo)resorcinol, 96%
C11H9N3O2
R0378
Reactive Red 120
C44H24Cl2N14Na6O20S6