EMAIL THIS PAGE TO A FRIEND
Journal of clinical rheumatology : practical reports on rheumatic & musculoskeletal diseases
A repeated oral administration study of febuxostat (TMX-67), a non-purine-selective inhibitor of xanthine oxidase, in patients with impaired renal function in Japan: pharmacokinetic and pharmacodynamic study.
PMID 21654267
Abstract
Allopurinol has been widely used for treatment of hyperuricemia, however, it may be associated with various adverse effects. Febuxostat has been identified as a potentially safe and efficacious alternative. A multicenter, open-label, parallel, between-group comparative study was conducted to investigate the effects of renal function on the pharmacokinetics, pharmacodynamics, and safety of febuxostat, a novel inhibitor of uric acid synthesis. Based on creatinine clearance (Ccr), 29 subjects were assigned to 3 groups: normal renal function (Ccr ≥ 80 mL/min), mild renal dysfunction (80 mL/min > Ccr ≥ 50 mL/min), or moderate renal dysfunction (50 mL/min > Ccr ≥ 30 mL/min). Febuxostat was repeatedly orally administered at a dose of 20 mg/d for 7 days. Impaired renal function caused a slight increase in systemic exposure to unchanged febuxostat and its oxidative metabolites, but the exposure did not increase through repeated administration. Moreover, renal impairment did not markedly reduce the effects of febuxostat on plasma uric acid levels. There were no clinically significant adverse events even in patients with impaired renal function. Febuxostat is considered an inhibitor of uric acid synthesis that could be used in patients with mild to moderate renal impairment without dose adjustment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O6881
Oxypurinol, ≥98% (HPLC)
C5H4N4O2