The Analyst
A novel electrochemiluminescence aptasensor for protein based on a sensitive N-(aminobutyl)-N-ethylisoluminol-functionalized gold nanoprobe.
PMID 21655607
A novel electrochemiluminescence (ECL) aptasensor for platelet-derived growth factor B chain (PDGF-BB) assay was developed by assembling N-(aminobutyl)-N-ethylisoluminol functionalized gold nanoparticles (ABEI-AuNPs) with aptamers as nanoprobes. In the protocol, the biotinylated aptamer capture probes were first immobilized on a streptavidin coated gold nanoparticle (AuNPs) modified electrode, afterwards, the target PDGF-BB and the ABEI-AuNPs tagged aptamer signal probe were successively attached to the modified electrode by virtue of the dimer structure of PDGF-BB to fabricate a "sandwich" conjugate modified electrode, i.e. an aptasensor. ECL measurement was carried out with a double-step potential in carbonate buffer solution containing H(2)O(2). The aptasensor showed high sensitivity and selectivity toward PDGF-BB and specificity toward PDGF-BB aptamer. The detection limit was as low as 2.7 × 10(-14) M. In this work, the ABEI-AuNPs synthesized by a simple seed growth method have been successfully used as aptamer labels, which greatly amplified the ECL signal by binding numbers of ABEI molecules on the surface of AuNPs. The ABEI-AuNPs signal amplification is superior to other reported signal amplification strategies based on aptamer-related polymerase chain reaction or functionalized nanoparticles in simplicity, stability, labeling property and practical applicability. And the ABEI-AuNPs based nanoprobe is more sensitive than the luminol functionalized AuNPs based nanoprobe. Moreover, such an ultra-sensitive and low-cost assay can be accomplished with a simple and fast procedure by using a simple ECL instrumentation. The aptasensor was also applied for the detection of PDGF-BB in human serum samples, showing great application potential. Given these advantages, the ECL aptasensor is well suited for the direct, sensitive and rapid detection of protein in complex clinical samples.
Related Materials
Product #
Molecular Formula
Add to Cart
N-(4-Aminobutyl)-N-ethylisoluminol, ≥90%