EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
High-performance liquid chromatographic determination and pharmacokinetic study of apigenin-7-O-β-D-glucoside in rat plasma after intravenous administration.
PMID 21656359
Abstract
The present study was designed to validate an analytical method based on HPLC for the quantitative measurement of the Apigenin-7-O-β-D-glucoside (AGL) in rat plasma after intravenous administration. HPLC analysis was done using a C18 column, UV detection at 335 nm, and a mobile phase of methanol:0.2% phosphoric acid (1:1, v/v). Good linearity was observed over the range of 0.06-7.20 μg/mL with a lower limit of quantication of 0.06 μg/mL. The intra-and inter-day precision values were below 9.97%. No interference peaks or matrix effects were observed. The method was fully validated and successfully applied to a pharmacokinetic study of AGL.
Related Materials