EMAIL THIS PAGE TO A FRIEND
ChemSusChem
Hydrogenation of p-chloronitrobenzene over nanostructured-carbon-supported ruthenium catalysts.
PMID 21656695
Abstract
Carbon nanotubes (CNTs) and carbon nanofibers (CNFs) have been used for the first time to support ruthenium nanoparticles for the hydrogenation of p-chloronitrobenzene (p-CNB) to produce selectively p-chloroaniline. The preparation of well-dispersed ruthenium catalysts from the [Ru(3)(CO)(12)] precursor required activation of the purified supports by nitric acid oxidation. The supports, purified and functionalized, and the supported catalysts have been characterized by a range of techniques. The catalytic activity of these materials for the hydrogenation of p-CNB at 35 bar and 60 °C is shown to reach as high as 18 mol(p-CNB)g(Ru)(-1) h(-1), which is one order of magnitude higher than a commercial Ru/Al(2)O(3) catalyst. Selectivities between 92 and 94 % are systematically obtained, the major byproduct being aniline.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45925
1-Chloro-4-nitrobenzene, analytical standard
C6H4ClNO2
C59122
1-Chloro-4-nitrobenzene, 99%
C6H4ClNO2