EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Biosensing platform based on fluorescence resonance energy transfer from upconverting nanocrystals to graphene oxide.
PMID 21656878