EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Separation of bases, phenols and pharmaceuticals on ionic liquid-modified silica stationary phase with pure water as mobile phase].
PMID 21657060
Abstract
N-methylimidazolium ionic liquid (IL) -modified silica was prepared with the reaction of 3-chloropropyl modified silica and N-methylimidazole using toluene as solvent. Based on the multiple interactions between N-methylimidazolium IL-modified silica and analytes such as hydrophobic interaction, electrostatic attraction, repulsion interaction, hydrogen-bonding, etc., the bases (cytosine, thymine, 2-aminopyrimidine and 6-chloroguanine), phenols (m-aminophenol, resorcinol and m-nitrophenol) and three pharmaceuticals (moroxydine hydrochloride, acyclovir and cephalexin hydrate) were separated successfully with only pure water as the mobile phase. These chromatographic separations are environmental friendly, economical and convenient, without any organic solvent or buffer additive. The retention mechanism of these samples on the stationary phase was also investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A78608
2-Aminopyrimidine, 97%
C4H5N3