EMAIL THIS PAGE TO A FRIEND
Evidence-based complementary and alternative medicine : eCAM
Traditional Japanese formula kigikenchuto accelerates healing of pressure-loading skin ulcer in rats.
PMID 21660308
Abstract
We evaluated the effect of kigikenchuto (KKT), a traditional Japanese formula, in a modified rat pressure-loading skin ulcer model. Rats were divided into three groups, KKT extract orally administered (250 or 500 mg/kg/day for 35 days) and control. KKT shortened the duration until healing. Immunohistochemically, KKT increased CD-31-positive vessels in early phase and increased α-smooth muscle actin-(α-SMA-) positive fibroblastic cells in early phase and decreased them in late phase of wound healing. By Western blotting, KKT showed the potential to decrease inflammatory cytokines (MCP-1, IL-1β, and TNF-α) in early phase, decrease vascular endothelial growth factor in early phase and increase it in late phase, and modulate the expression of extracellular protein matrix (α-SMA, TGF-β1, bFGF, collagen III, and collagen I). These results suggested the possibility that KKT accelerates pressure ulcer healing through decreases of inflammatory cytokines, increase of angiogenesis, and induction of extracellular matrix remodeling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295272
1,1-Difluoroethylene, ≥99%
C2H2F2