EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Facile fabrication of noble metal nanoparticles encapsulated in hollow silica with radially oriented mesopores: multiple roles of the N-lauroylsarcosine sodium surfactant.
PMID 21660319
Abstract
A facile one-pot method was reported to fabricate noble metal nanoparticles encapsulated in silica hollow nanospheres with radially oriented mesopores, and the anionic amino acid surfactant, N-lauroylsarcosine sodium, played multiple roles: reducing agent, stabilizer, emulsion droplets and mesopore template.
Related Materials