EMAIL THIS PAGE TO A FRIEND
Preparative biochemistry & biotechnology
Purification and characterization of pullulanase from Lactococcus lactis.
PMID 21660865
Abstract
This paper describes a simple and efficient method of isolation of a plullulanase type I from amylolytic lactic acid bacteria (ALAB). Extracellular pullulanase type I was purified from a cell-free culture supernatant of Lactococcus lactis IBB 500 by using ammonium sulfate fractionation and dialysis (instead of ultrafiltration), and ion-exchange chromatography with CM Sepharose FF followed by gel filtration chromatography with Sephadex G-150 as the final step. A final purification factor of 14.36 was achieved. The molecular mass of the enzyme was estimated as 73.9 kD. The optimum temperature for the enzyme activity was 45°C and the optimum pH was 4.5. Pullulanase activity was increased by addition Co(2+) and completely inhibited by Hg(2+). The enzyme activity was specifically directed toward α-1,6 glycosidic linkages of pullulan giving maltotriose units. Enzymatic hydrolysis of starch and amylose produced a mixture of maltose and maltotriose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E2412 Pullulanase microbial