EMAIL THIS PAGE TO A FRIEND
Hepato-gastroenterology
Chemoprevention of chemically-induced biliary carcinogenesis in hamsters by vitamin K2.
PMID 21661384
Abstract
Pancreaticobiliary maljunction (PBM) is a high risk factor of biliary tract cancer. The chemopreventive effects of Vitamin K2 (menaquinone-4: MK4) in a hamster PBM model were investigated. The extrahepatic bile duct at the distal end of the common duct was ligated and cholecystoduodenostomy was performed (Group I). The same surgery was performed and from 4 weeks after surgery, 10 mg/kg of N-nitrosobis (2-oxopropyl) amine was subcutaneously injected once a week with a one-week interval (Group II). In addition of Group II, MK4 was orally administered once a day, five times with every week (Group III). The hamsters were sacrificed 20 weeks after surgery and histopathological findings of gallbladder were investigated. Group I showed predominantly proper epithelium without cancer. In Group II, atypical epithelium (AE) was observed in 75% of animals and early cancer was observed in 25%. Group III showed less AE and no cancer. The PCNA labeling index in Group III was statistically significantly lower than in Group II. In addition, no statistically significant differences were noted among the groups in terms of the apoptosis labeling index. MK4 suppressed biliary carcinogenesis by the induction of cell cycle arrest in a hamster biliary carcinogenetic model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47774
Menaquinone (K2), analytical standard
C31H40O2
V9378
Vitamin K2
C31H40O2