EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Degradation studies of fenazaquin in soil under field and laboratory conditions.
PMID 21667312
Abstract
Degradation of fenazaquin in sandy loam soil was investigated under field and laboratory conditions. Fenazaquin (Magister 10EC) was applied @ 125 and 250 g a.i./ha in field and in pot under field capacity moisture in laboratory. Samples drawn periodically were analyzed on GC-NPD. The residues of fenazaquin in both the doses and conditions dissipated almost 90% in 90 days. Half-life period were 32.04 and 31.35 days at two doses, respectively at field conditions and 30.10 and 28.94 days at laboratory conditions. Dissipation was approximated to first order kinetics in both conditions having correlation coefficient ranging from -0.9848 to -0.9914.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31635
Fenazaquin, PESTANAL®, analytical standard
C20H22N2O