EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Entrapment and kinetic resolution of stabilized axial and equatorial conformers of spiro-β-lactams.
PMID 21667974
Abstract
The facile synthesis of the stabilized axial and equatorial conformers of spiro-β-lactams was achieved via entrapment of cyclohexanone imines (Schiff bases) with acetoxyacetyl chloride in a [2 + 2]-cycloaddition reaction followed by their kinetic resolution. The immobilization of the racemic substrates on an inert solid support significantly reduced the reaction time and improved the enantioselectivity of conformers during kinetic resolution. The mechanism of the formation of the spiro-β-lactams was explored using B3LYP/6-31+G* level quantum chemical calculations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
302368
Acetoxyacetyl chloride, 97%
C4H5ClO3