EMAIL THIS PAGE TO A FRIEND
Physical review letters
Scattering lens resolves sub-100 nm structures with visible light.
PMID 21668161
Abstract
The smallest structures that conventional lenses are able to optically resolve are of the order of 200 nm. We introduce a new type of lens that exploits multiple scattering of light to generate a scanning nanosized optical focus. With an experimental realization of this lens in gallium phosphide we imaged gold nanoparticles at 97 nm optical resolution. Our work is the first lens that provides a resolution better than 100 nm at visible wavelengths.