EMAIL THIS PAGE TO A FRIEND
Microbial pathogenesis
Interesting anticandidal effects of anisic aldehydes on growth and proton-pumping-ATPase-targeted activity.
PMID 21669279
Abstract
Attention has been drawn to evaluate the antifungal activity of p-anisaldehyde (1), o-anisaldehyde (2) and m-anisaldehyde (3). To put forward this approach, antifungal activity has been assessed in thirty six fluconazole-sensitive and eleven fluconazole-resistant Candida isolates. Growth and sensitivity of the organisms were significantly effected by test compounds at different concentrations. The rapid irreversible action of compound-1, compound-2 and compound-3 on fungal cells suggested a membrane-located target for their action. We investigated their effect on H(+) ATPase mediated H(+)-pumping by various Candida species. All the compounds inhibit H(+)- ATPase activity at their respective MIC(90) values. Inhibition of H(+) ATPase leads to intracellular acidification and cell death. Scanning electron microscopy analysis revealed deep wrinkles, deformity and flowed content. Furthermore, it was also observed that position of methoxy group attached to the benzene ring decides antifungal activity of the compound. The present study indicates that compound-1, compound-2 and compound-3 have significant antifungal activity against Candida, including azole-resistant strains, advocating further investigation for clinical applications in the treatment of fungal infections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
129658
m-Anisaldehyde, 97%
C8H8O2
97063
p-Anisaldehyde, analytical standard
C8H8O2