EMAIL THIS PAGE TO A FRIEND
Organic letters
Stereocontrolled synthesis of α-amino-α'-alkoxy ketones by a copper-catalyzed cross-coupling of peptidic thiol esters and α-alkoxyalkylstannanes.
PMID 21675755
Abstract
A stereocontrolled synthesis of α-amino-α'-alkoxy ketones is described. This pH-neutral copper(I) thiophene-2-carboxylate (CuTC)-catalyzed cross-coupling of amino acid thiol esters and chiral nonracemic α-alkoxyalkylstannanes gives α-amino-α'-alkoxy ketones in good to excellent yields with complete retention of configuration at the α-amino- and α-alkoxy-substituted stereocenters.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T32603
2-Thiophenecarboxylic acid, ReagentPlus®, 99%
C5H4O2S