EMAIL THIS PAGE TO A FRIEND
The Journal of neuropsychiatry and clinical neurosciences
The effect of riluzole augmentation in a patient with treatment-resistant obsessive-compulsive disorder, taking two other glutaminergic agents.
PMID 21677213
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6777
Phenelzine sulfate salt
C8H12N2 · H2O4S