EMAIL THIS PAGE TO A FRIEND
Parasite (Paris, France)
Sitamaquine as a putative antileishmanial drug candidate: from the mechanism of action to the risk of drug resistance.
PMID 21678786
Abstract
Sitamaquine is a 8-aminoquinoline in development for the treatment of visceral leishmaniasis by oral route, no activity being observed on the experimental cutaneous leishmaniasis experimental models. Recent data explain how sitamaquine accumulate in Leishmania parasites, however its molecular targets remain to be identified. An advantage of sitamaquine is its short elimination half-life, preventing a rapid resistance emergence. The antileishmanial action of its metabolites is not known. The selection of a sitamaquine-resistant clone of L. donovani in laboratory and the phase II clinical trials pointing out some adverse effects such as methemoglobinemia and nephrotoxicity are considered for a further development decision.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
260789
8-Aminoquinoline, 98%
C9H8N2
SML1542
Sitamaquine tosylate, ≥98% (HPLC)
C21H33N3O · C7H8O3S