EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Cutaneous lymphoid hyperplasia related to squaric acid dibutyl ester.
PMID 21679832
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
339792
3,4-Dibutoxy-3-cyclobutene-1,2-dione, 98%
C12H18O4