EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and anti-inflammatory effects of a series of novel 7-hydroxycoumarin derivatives.
PMID 21680063
Abstract
A number of 7-hydroxycoumarins have been synthesised by Pechmann cyclisation using differently substituted resorcinols employing perchloric acid as the condensing agent. All the compounds have been characterised by analytical and spectroscopic methods. The anti-inflammatory properties were tested with LPS-induced inflammation in J774 macrophages. Expression of iNOS and COX-2 was determined by Western blot, NO by nitrite assay and IL-6 by ELISA analyses. Fifteen of the tested 7-hydroxycoumarins also inhibited IL-6 production but none of them had any major inhibitory effect on COX-2 expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54826
Umbelliferone, analytical standard
C9H6O3