EMAIL THIS PAGE TO A FRIEND
The New Zealand medical journal
Hypoglycaemia after treatment with perhexiline maleate: a case report.
PMID 21681255
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0120
Perhexiline maleate salt, ≥98% (HPLC)
C19H35N · C4H4O4