EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
The two spin states of an end-on copper(II)-superoxide mimic.
PMID 21681293
Abstract
The reaction of nitrosobenzene with copper(I) complexes of a tetradentate ligand led to two novel species that are best described as copper(II) complexes of an O-bonded nitrosobenzyl radical anion, in either the singlet or the triplet spin-state. Both states were characterized by crystal structures, magnetic measurements and DFT calculations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N24609
Nitrosobenzene, ≥97%
C6H5NO