EMAIL THIS PAGE TO A FRIEND
Acta medica Iranica
A case report of unusual vasculitic reaction after Methocarbamol injection.
PMID 21681710
Abstract
Descriptive case report of a 42-year old woman with coetaneous vasculitis, and severe abdominal pain, which was led to diagnostic laparotomy. These presentations are probably as a side effect of Methocarbamol injection. This is the first report according to our literature search (PubMed, google scholar, ISI web of knowledge, ProQuest, MD consult, Science Direct, SCOPUS) about Methocarbamol related vasculitis from 1966 since now. Vasculitis is not a known side effect of Methocarbamol. This case indicates, likely the potential for development of vasculitis with this medication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7012
Guaiacol glyceryl ether carbamate, analytical standard
C11H15NO5