EMAIL THIS PAGE TO A FRIEND
Human mutation
N-carbamylglutamate enhancement of ureagenesis leads to discovery of a novel deleterious mutation in a newly defined enhancer of the NAGS gene and to effective therapy.
PMID 21681857
Abstract
N-acetylglutamate synthase (NAGS) catalyzes the conversion of glutamate and acetyl-CoA to NAG, the essential allosteric activator of carbamyl phosphate synthetase I, the first urea cycle enzyme in mammals. A 17-year-old female with recurrent hyperammonemia attacks, the cause of which remained undiagnosed for 8 years in spite of multiple molecular and biochemical investigations, showed markedly enhanced ureagenesis (measured by isotope incorporation) in response to N-carbamylglutamate (NCG). This led to sequencing of the regulatory regions of the NAGS gene and identification of a deleterious single-base substitution in the upstream enhancer. The homozygous mutation (c.-3064C>A), affecting a highly conserved nucleotide within the hepatic nuclear factor 1 (HNF-1) binding site, was not found in single nucleotide polymorphism databases and in a screen of 1,086 alleles from a diverse population. Functional assays demonstrated that this mutation decreases transcription and binding of HNF-1 to the NAGS gene, while a consensus HNF-1 binding sequence enhances binding to HNF-1 and increases transcription. Oral daily NCG therapy restored ureagenesis in this patient, normalizing her biochemical markers, and allowing discontinuation of alternate pathway therapy and normalization of her diet with no recurrence of hyperammonemia. Inc.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4375
N-Carbamyl-L-glutamic acid
C6H10N2O5