EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Discovery of a novel class of 2-aminopyrimidines as CDK1 and CDK2 inhibitors.
PMID 21684737
Abstract
A series of new 2-(2-aminopyrimidin-4-yl)phenol derivatives were synthesized as potential antitumor compounds. Substitution with pyrrolidine-3,4-diol at the 4-position of phenol provided potent inhibitory activity against CDK1 and CDK2. X-ray crystal structural studies were performed to account for the effect of the substituent on both the enzymatic and cell growth inhibitory activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A78608
2-Aminopyrimidine, 97%
C4H5N3