EMAIL THIS PAGE TO A FRIEND
Acta pharmaceutica (Zagreb, Croatia)
Compatibility studies of nateglinide with excipients in immediate release tablets.
PMID 21684850
Abstract
Experiments were done to assess the compatibility of nateglinide with selected excipients in the development of immediate release tablets of nateglinide by thermal and isothermal stress testing (IST) techniques. To evaluate the drug-excipient compatibility, different techniques such as differential scanning calorimetric (DSC) study, infra-red (IR) spectrophotometric study and isothermal stress testing were adopted. The results of DSC study showed that magnesium stearate exhibited some interaction with nateglinide. However, the results of IR, and IST studies showed that all the excipients used in the formula were compatible with nateglinide. Optimized formulations developed using the compatible excipients were found to be stable over 3 months of accelerated stability studies (40 ± 2°C and 75 ± 5% RH). Overall, compatibility of excipients with nateglinide was successfully evaluated using a combination of thermal and IST methods and the formulations developed using the compatible excipients were found to be stable.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3538
Nateglinide, ≥98% (HPLC), solid
C19H27NO3