EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Enhancing the photothermal stability of plasmonic metal nanoplates by a core-shell architecture.
PMID 21688329
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
148202
4-Mercaptopyridine, 95%
C5H5NS