EMAIL THIS PAGE TO A FRIEND
Mutagenesis
Effect of pretreatment with hepatic mixed-function oxidase inducers on the genotoxicity of four rat carcinogens in the hepatocyte/DNA repair assay.
PMID 2169015
Abstract
Four genotoxic rat carcinogens, previously reported as negative in the standard hepatocyte/DNA repair assay, were studied in hepatocyte cultures derived from the livers of rats pretreated with the hepatic mixed-function oxidase (MFO) inducers, Aroclor (ARO) and 3-methylcholanthrene (3-MC). Cytembena and dimethyl hydrogen phosphite induced positive dose-dependent DNA repair responses in ARO- and 3-MC-pretreated cultures, while 2,4-, 2,6-toluene diisocyanate, and ziram were negative for unscheduled DNA synthesis in cultures from both pretreatments. The results from this study indicate that pretreatment with hepatic MFO inducers lowers the false-negative detection rate of various genotoxic rat carcinogens and other organ-specific genotoxins and significantly increases the sensitivity of the in vitro rat hepatocyte/DNA repair assay.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41430
Dimethyl phosphite, purum, ≥98.0% (GC)
C2H7O3P
D178454
Dimethyl phosphite, 98%
C2H7O3P