EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Direct synthesis of nanoporous carbon nitride fibers using Al-based porous coordination polymers (Al-PCPs).
PMID 21691616
Abstract
We report a new synthetic route for preparation of nanoporous carbon nitride fibers with graphitic carbon nitride polymers, by calcination of Al-based porous coordination polymers (Al-PCPs) with dicyandiamide (DCDA) under a nitrogen atmosphere.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D76609
Dicyandiamide, 99%
C2H4N4