EMAIL THIS PAGE TO A FRIEND
The Medical journal of Australia
Lymphatic filariasis in Australia: an update on presentation, diagnosis and treatment.
PMID 21692728
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8765
Diethylcarbamazine citrate salt, ≥98%
C16H29N3O8