EMAIL THIS PAGE TO A FRIEND
PLoS neglected tropical diseases
Experimental transmission of Leishmania infantum by two major vectors: a comparison between a viscerotropic and a dermotropic strain.
PMID 21695108
Abstract
We quantified Leishmania infantum parasites transmitted by natural vectors for the first time. Both L. infantum strains studied, dermotropic CUK3 and viscerotropic IMT373, developed well in Phlebotomus perniciosus and Lutzomyia longipalpis. They produced heavy late-stage infection and colonized the stomodeal valve, which is a prerequisite for successful transmission. Infected sand fly females, and especially those that transmit parasites, feed significantly longer on the host (1.5-1.8 times) than non-transmitting females. Quantitative PCR revealed that P. perniciosus harboured more CUK3 strain parasites, while in L. longipalpis the intensity of infection was higher for the IMT373 strain. However, in both sand fly species the parasite load transmitted was higher for the strain with dermal tropism (CUK3). All but one sand fly female infected by the IMT373 strain transmitted less than 600 promastigotes; in contrast, 29% of L. longipalpis and 14% of P. perniciosus infected with the CUK3 strain transmitted more than 1000 parasites. The parasite number transmitted by individual sand flies ranged from 4 up to 4.19×10(4) promastigotes; thus, the maximal natural dose found was still about 250 times lower than the experimental challenge dose used in previous studies. This finding emphasizes the importance of determining the natural infective dose for the development of an accurate experimental model useful for the evaluation of new drugs and vaccines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0393 Medium 199, With Hanks′ salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
M5017 Medium 199, With Earle′s salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
M4530 Medium 199, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
M3769 Medium 199, Modified, with Earle′s salts, without L-glutamine, sodium bicarbonate, and phenol red, powder, suitable for cell culture
M2520 Medium 199, HEPES Modification, with Earle′s salts, L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
M0650 Medium 199, 10 ×, With Earle′s salts, without L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
M2154 Medium 199, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
M7528 Medium 199, HEPES Modification, with Earle′s salts, 25 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
M7653 Medium 199, With Hanks′ salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
M9163 Medium 199, 10 ×, With Hanks′ salts, without L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture