EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Matchstick-shaped Ag2S-ZnS heteronanostructures preserving both UV/blue and near-infrared photoluminescence.
PMID 21695752
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241474
Silver sulfide, 99.9% trace metals basis
Ag2S