PLoS ONE
Lipid-induced peroxidation in the intestine is involved in glucose homeostasis imbalance in mice.
PMID 21698161
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
611778
Chloroform solution, NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
613037
Chloroform solution, NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 4 mm × 8 in.
487163
Chloroform solution, NMR reference standard, 1% in acetone-d6 (99.9 atom % D)
487171
Chloroform solution, NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 8 mm × 8 in.
611743
Chloroform solution, NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
611786
Chloroform solution, NMR reference standard, 5% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
487759
Chloroform solution, NMR reference standard, 5% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
611859
Chloroform solution, NMR reference standard, 20% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
613304
Chloroform solution, NMR reference standard, 2% in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
613312
Chloroform solution, NMR reference standard, 2% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in.
48603
Chloroform solution, 200 μg/mL in methanol, analytical standard
40021
Chloroform solution, certified reference material, 5000 μg/mL in methanol
708976
Chloroform solution, NMR reference standard, 3% in acetone-d6 (99.9 atom % D)
717908
Chloroform solution, NMR reference standard, 50% in acetone-d6 (99.9 atom % D), chromium(III) acetylacetonate 0.2 %
733814
Chloroform solution, NMR reference standard, 10% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
720984
Chloroform solution, NMR reference standard, 0.3% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
720305
Chloroform solution, NMR reference standard, 3% in acetone-d6 (99.9 atom % D), TMS 0.2 %, NMR tube size 3 mm × 8 in.
720992
Chloroform solution, NMR reference standard, 0.3% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
717916
Chloroform solution, NMR reference standard, 5% in acetone-d6 (99.9 atom % D), chromium(III) acetylacetonate 0.2 %
735981
Chloroform solution, NMR reference standard, 2% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
736430
Chloroform solution, NMR reference standard, 5% in acetone-d6 (99.9 atom % D), chromium(III) acetylacetonate 2 %, NMR tube size 10 mm × 8 in.
736422
Chloroform solution, NMR reference standard, 50% in acetone-d6 (99.9 atom % D), chromium(III) acetylacetonate 0.2 %, NMR tube size 10 mm × 8 in.