EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
The cyclooxygenase-2 pathway via the PGE₂ EP2 receptor contributes to oligodendrocytes apoptosis in cuprizone-induced demyelination.
PMID 21699540
Abstract
Cyclooxygenases (COX)-1 and -2 are key enzymes required for the conversion of arachidonic acid to eicosanoids, potent mediators of inflammation. In patients with multiple sclerosis, COX-2 derived prostaglandins (PGs) are elevated in the CSF and COX-2 is up-regulated in demyelinating plaques. However, it is not known whether COX-2 activity contributes to oligodendrocyte death. In cuprizone-induced demyelination, oligodendrocyte apoptosis and a concomitant increase in the gene expression of COX-2 and PGE₂-EP2 receptor precede histological demyelination. COX-2 and EP2 receptor were expressed by oligodendrocytes, suggesting a causative role for the COX-2/EP2 pathway in the initiation of oligodendrocyte death and demyelination. COX-2 gene deletion, chronic treatment with the COX-2 selective inhibitor celecoxib, or with the EP2 receptor antagonist AH6809 reduced cuprizone-induced oligodendrocyte apoptosis, the degree of demyelination and motor dysfunction. These data indicate that the PGE₂ EP2 receptor contributes to oligodendrocyte apoptosis and open possible new therapeutic approaches for multiple sclerosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1221
AH6809, ≥98%, crystalline solid or supercooled liquid
C17H14O5