EMAIL THIS PAGE TO A FRIEND
Heart rhythm
Predicting the degree of drug-induced QT prolongation and the risk for torsades de pointes.
PMID 21699823
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9910
Ibutilide hemifumarate salt, ≥98% (HPLC)
C20H36N2O3S · 0.5C4H4O4